missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBL1Y Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA550884
Additional Details : Weight : 0.01000kg
Description
The antibody detects endogenous levels of total TBL1 protein.
TBL1Y Polyclonal antibody specifically detects TBL1Y in Human samples. It is validated for Immunohistochemistry (Paraffin).Specifications
TBL1Y | |
Polyclonal | |
Unconjugated | |
TBL1Y | |
F-box-like/WD repeat-containing protein TBL1Y; TBL1; TBL1Y; transducin (beta)-like 1, Y-linked; transducin beta-like 1; transducin beta-like protein 1Y; Transducin-beta-like protein 1, Y-linked | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
90665 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q9BQ87 | |
TBL1Y | |
Fusion protein of human transducin (beta)-like 1, Y-linked | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |