Läs mer
Invitrogen™ TBP Monoclonal Antibody (1TBP18)
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA1189
Produktkod. 15682195
  • 4375.00 SEK / 100µL
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
Beskrivning
Beskrivning
MA1-189 detects TATA-Binding Protein from human, mouse, and rat, canine, and non-human primate samples. TBP (human) has an approximate molecular weight of 38kDa, while other species' isoforms can range from 35-36kDa. MA1-189 has been successfully used in Western blot and immunofluorescence and flow cytometry applications. The MA1-189 immunogen is the N-terminal domain of the human TATA-binding protein (TBP).
RICH1 is a GTPase-activating protein (GAP). GAPs stimulate the intrinsic GTP hydrolysis of small G proteins, such as RHOA, RAC1, and CDC42.
TRUSTED_SUSTAINABILITY
Specifikationer
Specifikationer
TBP
Monoclonal
1 mg/mL
PBS with 1mg/mL BSA, 30% glycerol and 0.05% sodium azide; pH 7.4
P20226, P29037
Tbp
N-terminal domain of the human TATA-binding protein (TBP).
100 μL
Primary
Human, Mouse, Rat, Canine, Monkey
Antibody
IgG1
Flow Cytometry, Western Blot, Immunocytochemistry
1TBP18
Unconjugated
Tbp
Gtf2d; GTF2D1; HDL4; MGC117320; MGC126054; MGC126055; RP1-191N21.3; SCA17; TATA binding protein; TA-TA binding protein 1; TATA box binding protein; TATA sequence-binding protein; TATA-binding factor; TATA-binding protein; TATA-box binding protein; TATA-box binding protein N-terminal domain; TATA-box factor; tata-box-binding; TATA-box-binding protein; TBP; TBP0; TBP1; TF2D; TFIID; TFIID core protein; transcription initiation factor TFIID TBP subunit; Unknown (protein for MGC:138100)
Mouse
Protein A
RUO
117526, 21374, 611193, 6908
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Produktförslag
Produktförslag
Filmer
SDS
Dokument
Dokument
Certifikat
Kampanjer
Kampanjer