missing translation for 'onlineSavingsMsg'
Learn More

TBRG4 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 10177-2-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16886383

  • 1355.00 SEK / 20µL
Estimated Shipment: 31-03-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

TBRG4 (transforming growth factor beta regulator 4), also known as CPR2 (cell cycle progression restoration protein 2) or FAST 4 (FAST kinase domain-containing protein 4), is a 631 amino acid protein that contains one RAP domain and belongs to the FAST kinase family. TBRG4 is ubiquitously expressed and may have a role in cell cycle progression. Existing as two alternatively spliced isoforms, the gene encoding TBRG4 maps to human chromosome 7p13.
Specifications

Specifications

TBRG4
Polyclonal
Unconjugated
TBRG4
CPR2, FASTKD4, KIAA0948, Protein TBRG4, TBRG4
Rabbit
Antigen Affinity Chromatography
RUO
9238
-20°C
Liquid
Western Blot
0.62 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q969Z0
TBRG4
TBRG4 Fusion Protein Ag0212
20 μL
Primary
Human
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers