Learn More
TBS with Tween™ (TBST), 20X Solution, Molecular Biology Grade, Ultrapure
Product Code. 15815428 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 L
5 L
Unit Size:
1 L
5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15815428 5 L 5 L
15805428 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15815428 Supplier Thermo Scientific Supplier No. J77500.K8
  • ONLINE EXCLUSIVE PRICE
    4845.00 SEK
    3996.65 SEK / 5 L
    Save 848.35 SEK
    17% Av
 Restricted Item
I lager
Add to basket
Edge
Used in Western blotting protocols
Used in Western blotting protocols.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material TBS with Tween (TBST)
Concentration 20X
Physical Form Solution
Quantity 5 L