Learn More
TBX2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA552582
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: IPMPTFGGLF PYPYTYMAAA AAAASALPAT SAAAAAAAAA GSLSRSPFLG SARPRLRFSP YQIPVTIPPS TSLLTTGLAS EGSKAAGGNS REPSPLPELA LRKVGAPSRG ALSPSGSAKE AANELQSIQR LVSGL Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 49%.
TBX2 Polyclonal antibody specifically detects TBX2 in Human samples. It is validated for Immunocytochemistry.Specifications
TBX2 | |
Polyclonal | |
Unconjugated | |
TBX2 | |
FLJ10169; LOW QUALITY PROTEIN: T-box transcription factor TBX2; T-box 2; T-box 2 protein; T-box 2 transcription factor; T-box protein 2; T-box transcription factor 2; T-box transcription factor TBX2; TBX2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6909 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q13207 | |
TBX2 | |
Recombinant protein corresponding to Human TBX2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only