missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBX6 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA567523
Additional Details : Weight : 0.01000kg
Description
TBX6 Polyclonal antibody specifically detects TBX6 in Human samples. It is validated for Immunohistochemistry (Paraffin).Specifications
TBX6 | |
Polyclonal | |
Unconjugated | |
TBX6 | |
rib-vertebrae; rv; SCDO5; T-box 6; T-box protein 6; T-box transcription factor 6; T-box transcription factor TBX6; TBX6 | |
Rabbit | |
affinity chromatography | |
RUO | |
6911 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
O95947 | |
TBX6 | |
Fusion protein of human TBX6 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |