Läs mer
TC E3 5031, Tocris Bioscience™
GREENER_CHOICE
Carboxylic acid-functionalized Thalidomide; cereblon ligand for PROTACs
Brand:  Tocris Bioscience 6466/100
Encompass
Produktkod. 15963091
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter »
Beskrivning
Beskrivning
TC E3 5031 is a derivative of Thalidomide (Cat.No. 0652), commonly used as a precursor to PROTAC® that hijacks cereblon as the E3 ubiquitin ligase component. Supplied with a carboxylic acid functional handle at a position known not to significantly affect binding to cereblon, for ready conjugation to a linker/target protein ligand.
PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
Require a different derivative? Get in touch .
Specifikationer
Specifikationer
1061605-21-7
2-[[2-(2,6-Dioxo-3-piperidinyl)-2,3-dihydro-1,3-dioxo-1H-isoindol-4-yl]oxy]acetic acid