Läs mer
Tocris Bioscience™ TC-F 2
GREENER_CHOICE
Produktkod. 18034992
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18034992 10 mg
18014262 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18034992 Leverantör Tocris Bioscience™ Leverantörsnummer 4355/10
Potent, reversible and selective FAAH inhibitor
Potent, reversible inhibitor of fatty acid amide hydrolase (FAAH) (IC50 values are 28 and 100 nM for human and rat FAAH respectively). Noncovalent inhibitor; displays selectivity for FAAH over cannabinoid-related targets (IC50 > 20 μM for CB1, CB2 and TRPV1). Active in vivo.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1304778-15-1
Molekylformel C26H25N5O2
Specifikationer
Kemiskt namn eller material TC-F 2
Kvantitet 10 mg
Formel vikt 439.51
Renhet >99%
Molekylvikt (g/mol) 439.519