Läs mer
TC JL 37, Tocris Bioscience™
Potent TYK2 inhibitor
Brand: Tocris Bioscience 6125/10
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
TC JL 37 is a potent inhibitor of tyrosine kinase 2 (TYK2, Ki = 1.6 nM). Shows some selectivity for TYK2 over JAK1/2/3. Inhibits IFNγ production in vivo in mice. Orally bioavailable.Specifikationer
| 2-Chloro-4-cyano-6-fluoro-N-[2-[[[(1R,2R)-2-fluorocyclopropyl]carbonyl]amino]-4-pyridinyl]benzamide | |
| 1258294-34-6 | |
| >98% | |
| 10 mg |