Läs mer
Tocris Bioscience™ TC LPA5 4
GREENER_CHOICE
Produktkod. 30001664
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30001664 10 mg
30001519 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30001664 Leverantör Tocris Bioscience™ Leverantörsnummer 4708/10
LPA5 receptor antagonist
TC LPA5 4 is an LPA5 receptor antagonist (IC50 = 0.8 μM in LPA5-RH7777 cells). TC LPA5 4 inhibits LPA-induced aggregation of isolated human platelets. Exhibits selectivity for LPA5 against 80 other screened targets.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1393814-38-4
Molekylformel C23H23ClN2O3
Specifikationer
Kemiskt namn eller material TC LPA5 4
Kvantitet 10 mg
Formel vikt 410.89
Renhet >98%
Molekylvikt (g/mol) 410.898