Läs mer
Tocris Bioscience™ TC-S 7009
GREENER_CHOICE
Produktkod. 18747418
Change view
Kvantitet:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18747418 25 mg
18771488 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18747418 Leverantör Tocris Bioscience™ Leverantörsnummer 5243/25
High affinity and selective HIF-2α inhibitor
TC-S 7009 is a high affinity and selective HIF-2α inhibitor (Kd = 81 nM). Binds to the HIF-2α PAS-B domain; disrupts HIF-2α-ARNT heterodimerization. Exhibits >60-fold selectivity for HIF-2α over HIF-1α. Decreases HIF-2α DNA-binding and attenuates expression of HIF-2α target genes in vitro.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-(3-Chloro-5-fluorophenyl)-4-nitro-2,1,3-benzoxadiazol-5-amine
CAS 1422955-31-4
Analysprocentintervall >99%
Kvantitet 25 mg
Molekylformel C12H6ClFN4O3
Formel vikt Observed MW: 308.65