Learn More
HUBER TC100E Immersion Chiller
For many applications immersion coolers are also an environmentally friendly alternative to mains water cooling. The TC immersion coolers are suitable for unsupervised use, for cooling tasks which require continuous cooling or as a variant with temperature control and a Pt100 probe.
Brand: HUBER 3005.0044.99
Description
- TC100E Immersion cooler, with natural refrigerant
- Rapid cooling of fluids
- Ideal for counter-cooling or as a replacement of dry ice
- LED temperature display
- Easy operation with only three eys
- Compact, space-saving construction
- Robust housing with carrying grips
- High quality stainless steel immersion probe
- Temperature range: -100°C to 40°C
- Cooling capacity (-80°C): 0.12kW
- Cooling capacity (0°C): 0.16kW
Specifications
-100°C to +40°C | |
TC100E |
|
230 V |