Läs mer
Tocris Bioscience™ TCB-2
Beskrivning
TCB-2 is a high affinity 5-HT2A receptor agonist (Ki values are 0.73 and 0.75 nM for rat and human receptors respectively). Potently stimulates IP3 accumulation in NIH3T3 cells stably expressing rat 5-HT2A receptors (EC50 = 36 nM). Induces head twitches and hypothermia in mice following i.p. administration.
Specifikationer
Specifikationer
| Kemiskt namn eller material | (4-Bromo-3,6-dimethoxybenzocyclobuten-1-yl)methylamine hydrobromide |
| CAS | 912342-28-0 |
| Analysprocentintervall | >99% |
| Kvantitet | 10 mg |
| Löslighet | Soluble to 100mM in DMSO |
| Molekylformel | C11H14BrNO2.HBr |
| Formel vikt | Observed MW: 353.05 |
For Research Use Only