missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ TCEA1 (Human) Recombinant Protein
Human TCEA1 full-length ORF ( NP_006747.1, 1 a.a. - 301 a.a.) recombinant protein with GST-tag at N-terminal.
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_006747.1 |
---|---|
Gene ID (Entrez) | 6917 |
Name | transcription elongation factor A (SII), 1 |
Preparation Method | Wheat germ expression system |
Quality Control Testing | 125% SDS-PAGE Stained with Coomassie Blue |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16181232
|
Abnova™
H00006917-P01.10UG |
10 μg |
4515.00 SEK
10µg |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16191232
|
Abnova™
H00006917-P01.25UG |
25 μg |
6845.00 SEK
25µg |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Sequence: MEDEVVRFAKKMDKMVQKKNAAGALDLLKELKNIPMTLELLQSTRIGMSVNAIRKQSTDEEVTSLAKSLIKSWKKLLDGPSTEKDLDEKKKEPAITSQNSPEAREESTSSGNVSNRKDETNARDTYVSSFPRAPSTSDSVRLKCREMLAAALRTGDDYIAIGADEEELGSQIEEAIYQEIRNTDMKYKNRVRSRISNLKDAKNPNLRKNVLCGNIPPDLFARMTAEEMASDELKEMRKNLTKEAIREHQMAKTGGTQTDLFTCGKCKKKNCTYTQVQTRSADEPMTTFVVCNECGNRWKFC
Specifications
NP_006747.1 | |
transcription elongation factor A (SII), 1 | |
125% SDS-PAGE Stained with Coomassie Blue | |
GTF2S, SII, TCEA, TF2S, TFIIS | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |
6917 | |
Wheat germ expression system | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
TCEA1 | |
GST |