missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ TCEA1 (Human) Recombinant Protein

Human TCEA1 full-length ORF ( NP_006747.1, 1 a.a. - 301 a.a.) recombinant protein with GST-tag at N-terminal.

Brand:  Abnova™ H00006917-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16181232

  • 3343.96 SEK / 10µg

Estimated Shipment: 09-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Sequence: MEDEVVRFAKKMDKMVQKKNAAGALDLLKELKNIPMTLELLQSTRIGMSVNAIRKQSTDEEVTSLAKSLIKSWKKLLDGPSTEKDLDEKKKEPAITSQNSPEAREESTSSGNVSNRKDETNARDTYVSSFPRAPSTSDSVRLKCREMLAAALRTGDDYIAIGADEEELGSQIEEAIYQEIRNTDMKYKNRVRSRISNLKDAKNPNLRKNVLCGNIPPDLFARMTAEEMASDELKEMRKNLTKEAIREHQMAKTGGTQTDLFTCGKCKKKNCTYTQVQTRSADEPMTTFVVCNECGNRWKFC
Specifications

Specifications

NP_006747.1
transcription elongation factor A (SII), 1
125% SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
TCEA1
GST
6917
Wheat germ expression system
10 μg
GTF2S, SII, TCEA, TF2S, TFIIS
Wheat Germ (in vitro)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
Videos
SDS
Documents
Certificates