missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCEA3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17158-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Necessary for efficient RNA polymerase II transcription elongation past template-encoded arresting sites. The arresting sites in DNA have the property of trapping a certain fraction of elongating RNA polymerases that pass through, resulting in locked ternary complexes. Cleavage of the nascent transcript by S-II allows the resumption of elongation from the new 3′-terminus.Specifications
TCEA3 | |
Polyclonal | |
Unconjugated | |
TCEA3 | |
TCEA3, TFIIS, TFIIS.H, TFIISH | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
21401, 298559, 6920 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O75764, P23881 | |
TCEA3 | |
TCEA3 Fusion Protein Ag10404 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |