missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCEAL3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA561034
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: PKDSQEDLQE RHLSSEEMMR ECGDVSRAQE ELRK Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 79%.
TCEAL3 encodes a member of the transcription elongation factor A (SII)-like (TCEAL) gene family. Members of this family contain TFA domains and may function as nuclear phosphoproteins that modulate transcription in a promoter context-dependent manner. Multiple family members are located on the X chromosome. Alternative splicing results in multiple transcript variants encoding a single isoform.Specifications
TCEAL3 | |
Polyclonal | |
Unconjugated | |
TCEAL3 | |
1100001D19Rik; MSTP072; TCEAL3; TCEA-like protein 3; transcription elongation factor A (SII)-like 3; transcription elongation factor A like 3; transcription elongation factor A protein-like 3; Transcription elongation factor S-II protein-like 3; WEX8 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
85012 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q969E4 | |
TCEAL3 | |
Recombinant protein corresponding to Human TCEAL3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only