missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCEB1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA550802
Additional Details : Weight : 0.01000kg
Description
The antibody detects endogenous levels of total TCEB1 protein.
TCEB1 Polyclonal antibody specifically detects TCEB1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin).Specifications
TCEB1 | |
Polyclonal | |
Unconjugated | |
ELOC | |
2610043E24Rik; 2610301I15Rik; AA407206; AI987979; AW049146; eloC; elongation factor SIII p15 subunit; elongin 15 kDa subunit; elongin C; elongin-C; RNA polymerase II transcription factor SIII subunit C; SIII; SIII p15; Stromal membrane-associated protein SMAP1B homolog; Tceb1; transcription elongation factor B (SIII), polypeptide 1; transcription elongation factor B (SIII), polypeptide 1 (15kDa, elongin C); transcription elongation factor B polypeptide 1; transcription elongation factor B polypeptide 1; elongin-C; transcription elongation factor B subunit 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
64525, 67923, 6921 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
2.2 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
P83940, P83941, Q15369 | |
ELOC | |
Full length fusion protein of Human TCEB1 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |