Learn More
Thermo Scientific™ TCF-32 Continuous-Flow Rotors
Product Code. 12893412 Shop All Thermo Scientific Products
Change view
Click to view available options
Capacity:
940 mL
940mL/430 mL
Description:
940mL Accessory Core
TCF-32 Continuous-Flow Rotor
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity Description Unit Size
12893412 940 mL 940mL Accessory Core Each
12141670 940mL/430 mL TCF-32 Continuous-Flow Rotor Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12893412 Supplier Thermo Scientific™ Supplier No. 45830
  • ONLINE EXCLUSIVE PRICE
    482520.74 SEK
    353558.56 SEK / Paket med 1
    Save 128962.18 SEK
    26% Av
 Restricted Item
Beräknad leverans: 11-03-2027
Perform multi-liter processing applications that require high RCFs such as viral separations, particle isolations and vaccine production with the help of these rotors.
Perform multi-liter processing applications that require high RCFs such as viral separations, particle isolations and vaccine production with the help of Thermo Scientific™ TCF-32 Continuous-Flow Rotors.
Compatible with:
Thermo Scientific Sorvall WX Ultra Centrifuge Series
Ordering Information:
Core is required for operation. Must be purchased separately.
TRUSTED_SUSTAINABILITY
Specifications
Description 940mL Accessory Core
Type Continuous Flow Rotor
Capacity 940 mL
Max. RCF 102,000 x g
Max. Speed 32,000 rpm
K Factor 106
For Use With (Equipment) Thermo Scientific™ Sorvall™ WX ultraCentrifuges