Learn More
TCF12 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA566799
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: FSPPVNSGKTR PTTLGSSQFS GSGIDERGGT TSWGTSGQPS PSYDSSRGFT DSPHYSDHLN DSRLGAHEGL SPTPFMNSNL MGKTSERGSF SLYSRDTGLP GCQSSLLRQD LGLGSPAQLS SSGKPGTAYY SFSATSSRRR P Highest antigen sequence identity to the following orthologs - mouse 96%, rat 96%.
TCF12 Polyclonal antibody specifically detects TCF12 in Human samples. It is validated for Immunocytochemistry.Specifications
TCF12 | |
Polyclonal | |
Unconjugated | |
TCF12 | |
A130037E08Rik; ALF1; bHLHb20; Class A helix-loop-helix transcription factor ME1; class B basic helix-loop-helix protein 20; CRS3; DNA-binding protein HTF4; E-box-binding protein; HEB; HEBAlt; helix-loop-helix transcription factor 4; HsT17266; HTF4; HTF-4; ME1; Meb; REB; Salivary-specific cAMP response element-binding protein alpha; SCBP alpha; SCBPA; SCBP-alpha; TCF12; TCF-12; Transcription factor 12; transcription factor HTF-4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6938 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q99081 | |
TCF12 | |
Recombinant Human TCF12 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only