missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCF15 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA566313
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: CLSNQRKGGGR RDLGGSCLKV RGVAPLRGPR R Highest antigen sequence identity to the following orthologs - mouse 97%, rat 97%.
TCF15 is found in the nucleus and may be involved in the early transcriptional regulation of patterning of the mesoderm. The basic helix-loop-helix protein requires dimerization with another basic helix-loop-helix protein for efficient DNA binding.The protein encoded by this gene is found in the nucleus and may be involved in the early transcriptional regulation of patterning of the mesoderm. The encoded basic helix-loop-helix protein requires dimerization with another basic helix-loop-helix protein for efficient DNA binding.Specifications
TCF15 | |
Polyclonal | |
Unconjugated | |
TCF15 | |
basic helix-loop-helix transcription factor 15; bHLHa40; Bhlhec2; bHLH-EC2; Class A basic helix-loop-helix protein 40; EC2; LOW QUALITY PROTEIN: transcription factor 15; Meso1; par1; par1 protein; paraxis; Protein bHLH-EC2; TCF15; TCF-15; Transcription factor 15; transcription factor 15 (basic helix-loop-helix) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6939 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q12870 | |
TCF15 | |
Recombinant Human TCF15 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only