missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ TCF7L1 Polyclonal Antibody

Beskrivning
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human TCF7L1.
This gene encodes a member of the T cell factor/lymphoid enhancer factor family of transcription factors. These transcription factors are activated by beta catenin, mediate the Wnt signaling pathway and are antagonized by the transforming growth factor beta signaling pathway. The encoded protein contains a high mobility group-box DNA binding domain and participates in the regulation of cell cycle genes and cellular senescence.
Specifikationer
Specifikationer
| Antigen | TCF7L1 |
| Användningsområden | Immunoprecipitation, Western Blot |
| Klassificering | Polyclonal |
| Koncentration | 1 mg/mL |
| Konjugera | Unconjugated |
| Formulering | PBS with no preservative |
| Gen | TCF7L1 |
| Genaccessionsnr. | Q9HCS4 |
| Gene Alias | bHLHb21; HMG box transcription factor 3; T-cell factor 3; Tcf3; TCF-3; TCF7L1; transcription factor 3; transcription factor 7 like 1; transcription factor 7 like 1 (T cell specific, HMG box); transcription factor 7-like 1; transcription factor 7-like 1 (T-cell specific, HMG box); transcription factor 7-like 1 (T-cell specific, HMG-box); transcription factor 7-like 2 (T-cell specific, HMG box) |
| Gensymboler | TCF7L1 |
| Visa mer |
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?