missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCHP Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA558475
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: MEAFRQKAEL GRFLRHQYNA QLSRRTQQIQ EELEADRRIL QALLEKEDES QRLHLARREQ VMADVAWMKQ AIEEQ Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 76%.
TCHP Polyclonal antibody specifically detects TCHP in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Specifications
TCHP | |
Polyclonal | |
Unconjugated | |
TCHP | |
A930031F18Rik; AV279199; Mitochondrial protein with oncostatic activity; mitostatin; Protein TCHP; Tchp; TpMs; trichoplein keratin filament binding; trichoplein keratin filament-binding protein; trichoplein, keratin filament binding; tumor suppressor protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84260 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.5 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BT92 | |
TCHP | |
Recombinant protein corresponding to Human TCHP | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only