missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCP10 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14538-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
The exact function of TCP10 remains unknown.Specifications
TCP10 | |
Polyclonal | |
Unconjugated | |
TCP10 | |
t complex 10 homolog (mouse), T complex protein 10A homolog, TCP10, TCP10A | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
21458, 6953 | |
-20°C | |
Liquid |
Immunofluorescence, Immunocytochemistry, Western Blot | |
0.52 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q12799 | |
TCP10 | |
TCP10 Fusion Protein Ag6041 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |