missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
TCR beta Antibody (H57-597), Alexa Fluor™ 647 , Novus Biologicals™
Armenian Hamster Monoclonal Antibody
Brand: Novus Biologicals NBP3-20117AF647
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
TCR beta Monoclonal antibody specifically detects TCR beta in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry
Specifikationer
| TCR beta | |
| Monoclonal | |
| Alexa Fluor 647 | |
| T cell receptor beta chain, TCRB, TCRbeta, Tib | |
| Hamsters were immunised with affinity purified DO-11.10 TCR, and produced detectable anti-TCR antibodies which were judged by the ability of the anti-sera to immunoprecipitate TCR from surface labelled DO-11.10 T cell hybridomas. Hamster 57 was selected for hybridisation, and B cell hybridomas were produced using polyethylene glycol fusion of hamster splenic B cells to the mouse myeloma variant. Selected mAb were screened after subsequent culturing for their ability to immunoprecipitate TCR from detergent lysates of surface-radiolabelled murine T cell hybridomas. | |
| 0.1 mL | |
| Adaptive Immunity, Cell Biology, Diabetes Research, Immunology, Protein Kinase, Signal Transduction, Tyrosine Kinases | |
| 21577 | |
| Store at 4°C in the dark. | |
| IgG λ |
| Flow Cytometry, Immunohistochemistry | |
| H57-597 | |
| 50mM Sodium Borate | |
| Armenian Hamster | |
| Protein A purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering