missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCR gamma/delta Monoclonal Antibody (5A6.E9), FITC, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MHGD014
Additional Details : Weight : 0.50000kg
Description
Commonly used, FITC conjugates provide relatively high absorptivity, excellent fluorescence quantum yield, and good water solubility.
TCR gamma/delta Monoclonal antibody specifically detects TCR gamma/delta in Human samples. It is validated for Flow Cytometry.Specifications
TCR gamma/delta | |
Monoclonal | |
FITC | |
TRD | |
RGD1311300; similar to T cell receptor V delta 6; T cell receptor gamma locus; T3/TCR complex; TCR delta gamma; TCR gamma/delta; TCR-CD3 complex; Tcrg; Trg; Trg@ | |
Mouse | |
purified | |
RUO, ASR/IVD | |
6964, 6965 | |
4° C, store in dark | |
Liquid |
Flow Cytometry | |
5A6.E9 | |
PBS with 4mg/mL BSA and 0.1% sodium azide | |
0 | |
TRD, Trg | |
Human TCR gamma/delta T cell receptor | |
2 mL | |
Primary | |
Human | |
Antibody | |
IgG1 |