missing translation for 'onlineSavingsMsg'
Läs mer

TCR V beta 5b Monoclonal Antibody (W112), Invitrogen™

Produktkod. 11828831
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
200 μg
Förpackningsstorlek:
200µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
11828831 200 μg 200µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 11828831 Leverantör Invitrogen Leverantörsnummer TCR1645

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter

Denna artikel kan inte returneras. Se returpolicy

Mouse Monoclonal Antibody

TCR1645 targets TCR V beta 5b in FACS, IHC (F), IP, TCA, and WB applications and shows reactivity with Human samples. The TCR1645 immunogen is human TCR Vb5.3.

The ability of T cell receptors (TCR) to discriminate foreign from self-peptides presented by major histocompatibility complex (MHC) class II molecules is essential for an effective adaptive immune response. TCR recognition of self-peptides has been linked to autoimmune disease. Mutant self-peptides have been associated with tumors. Engagement of TCRs by a family of bacterial toxins know as superantigens has been responsible for toxic shock syndrome. Autoantibodies to V beta segments of T cell receptors have been isolated from patients with rheumatoid arthritis (RA) and systemic lupus erythematosus (SLE). The autoantibodies block TH1-mediated inflammatory autodestructive reactions and are believed to be a method by which the immune system compensates for disease (ref5). T Cell and TCR Diversity Most human T cells express the TCR alpha-beta and either CD4 or CD8 molecule (single positive, SP). A small number of T cells lack both CD4 and CD8 (double negative, DN). Increased percentages of alpha-beta DN T cells have been identified in some autoimmune and immunodeficiency disorders. Gamma-delta T cells are primarily found within the epithelium. They show less TCR diversity and recognize antigens differently than alpha-beta T cells. Subsets of gamma-delta T cells have shown antitumor and immunoregulatory activity.
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen TCR V beta 5b
Användningsområden Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation, T-Cell Activation, Western Blot
Klassificering Monoclonal
Klona W112
Koncentration 0.15 mg/mL
Konjugera Unconjugated
Formulering PBS with 0.5% BSA and 0.1% sodium azide
Gen TRB
Genaccessionsnr. 0
Gene Alias T cell receptor beta locus; TCRB; TRB; TRB@
Gensymboler TRB
Värd art Mouse
Immunogen Human TCR Vb5.3
Reningsmetod Protein G
Kvantitet 200 μg
Regulatorisk status RUO
Primär eller sekundär Primary
Gen-ID (Entrez) 6957
Målarter Human
Innehåll och lagring -20°C
Produkttyp Antibody
Form Liquid
Isotyp IgG1
Visa mer Visa mindre
Produkttitel
Välj ett problem

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.