Learn More
Invitrogen™ Human TCTN2 Synthetic Peptide
Synthetic Peptide
Brand: Invitrogen™ PEP1098
Additional Details : Weight : 0.01000kg
Description
TCTN2 Synthetic Peptide, PEP-1098, from Invitrogen.
This peptide corresponds to 15 amino acids near the carboxy terminus of human TCTN2.PEP-1098 can be used as a blocking peptide with polyclonal antibody PA5-20984.Specifications
79867 | |
purified | |
Human TCTN2 | |
-20°C | |
Liquid | |
Blocking Assay, Control | |
No | |
Peptide | |
RUO - research use only |
Human | |
Q96GX1 | |
200 μg/mL | |
Synthetic Peptide | |
PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 | |
TCTN2 | |
0.02% sodium azide | |
50 μg | |
PBS |