Läs mer
Tocris Bioscience™ TDI 11861
Beskrivning
TDI 11861 is a potent and selective soluble adenylyl cyclase (sAC) inhibitor (IC50 values are 3.3 nM and 5.5 nM in biochemical and cellular assays, respectively; Kd = 1.4 nM). Inhibits cellular sAC-specific cAMP accumulation (IC50 = 7 nM) and blocks sperm capacitation in vitro. Cell permeable, not cytotoxic and suitable for both in vivo and in vitro use.
Specifikationer
Specifikationer
| Kemiskt namn eller material | (3R)-4-[2-[2-[[3-(2-Amino-6-chloro-4-pyrimidinyl)-1-(difluoromethyl)-1H-pyrazol-4-yl]methyl]phenoxy]ethyl]-3-morpholinemethanol |
| CAS | 2857049-72-8 |
| Kvantitet | 5 mg |
| Mål | Adenylyl Cyclase Inhibitors |
| Molekylformel | C22H25ClF2N6O3 |
| Renhet | 98% |