Learn More
TDRD10 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA565861
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: VMFIDFGQLAT IPVQSLRSLD SDDFWTIPPL TQPFMLEKDI LSSYEVVHRI LKGKITGALN SAVTAPASNL AVVPPLL Highest antigen sequence identity to the following orthologs - mouse 26%, rat 24%.
TDRD10 gene ontology annotations related to this gene include RNA binding.Specifications
TDRD10 | |
Polyclonal | |
Unconjugated | |
TDRD10 | |
TDRD10; tudor domain containing 10; tudor domain-containing protein 10; UNQ9380/PRO34205 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
126668 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5VZ19 | |
TDRD10 | |
Recombinant Human TDRD10 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only