missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TDRD5 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | TDRD5 |
---|---|
Concentration | 0.39 mg/mL |
Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16823815
|
Proteintech
25239-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 22-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16813815
|
Proteintech
25239-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 22-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
TDRD5, also named as Tudor domain-containing protein 5, is a 981 amino acid protein, which contains one Tudor domain and three HTH OST-type domains. TDRD5 localizes in the cytoplasm and belongs to the TDRD5 family. TDRD5 is required during spermiogenesis to participate in the repression transposable elements and prevent their mobilization, which is essential for the germline integrity.Specifications
TDRD5 | |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q5VCS6, Q8NAT2 | |
163589, 214575 | |
TDRD5 Fusion Protein Ag18896 | |
Primary | |
-20°C | |
TDRD5 |
0.39 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
RP11 427G13.1, TDRD5, tudor domain containing 5, TUDOR3 | |
Tdrd5 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |