missing translation for 'onlineSavingsMsg'
Learn More

TE buffer, (20X), pH 7.6, autoclaved, Thermo Scientific™

Catalog Number p-7076564
Click to view available options
Quantity:
250 mL
500 mL
This item is not returnable. View return policy
1185-53-1
C4H12ClNO3
157.594
MFCD00236359
QKNYBSVHEMOAJP-UHFFFAOYSA-N
93573
2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride
C(C(CO)(CO)N)O.Cl
This item is not returnable. View return policy
1185-53-1
C4H12ClNO3
157.594
MFCD00236359
QKNYBSVHEMOAJP-UHFFFAOYSA-N
93573
2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride
C(C(CO)(CO)N)O.Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Physical Form Liquid
Quantity 250 mL
Packaging Plastic bottle
Chemical Name or Material TE buffer
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'