missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TE buffer, pH 7.4, RNAse free, Thermo Scientific Chemicals
Click to view available options
Quantity:
125 mL
250 mL
Description
TE buffer is a commonly used buffer solution in procedures involving DNA or RNA. It is used for storing and diluting DNA and RNA. Treatment with TE buffer enhances the staining intensity of antibodies in the immunohistochemical detection of certain proteins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
TE buffer is a commonly used buffer solution in procedures involving DNA or RNA. It is used for storing and diluting DNA and RNA. Treatment with TE buffer enhances the staining intensity of antibodies in the immunohistochemical detection of certain proteins.
Solubility
Fully miscible in water.
Notes
200mM Tris-HCl with 20mM EDTA
TE buffer is a commonly used buffer solution in procedures involving DNA or RNA. It is used for storing and diluting DNA and RNA. Treatment with TE buffer enhances the staining intensity of antibodies in the immunohistochemical detection of certain proteins.
Solubility
Fully miscible in water.
Notes
200mM Tris-HCl with 20mM EDTA
Specifications
Color | Undesignated |
pH | 7.4 |
Physical Form | Liquid |
Quantity | 125 mL |
Solubility Information | Fully miscible in water. |
Chemical Name or Material | TE buffer, RNAse free |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'