Läs mer
Tebipenem, MedChemExpress
Produktkod. 16488288
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16488288 10 mg
16474177 50 mg
16420728 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 16488288 Leverantör Medchem Express Leverantörsnummer HYA0076
MedChemExpress Tebipenem is an orally available carbapenem antibiotic, shows broad-spectrum activity against Gram-positive and -negative bacteria, except for Pseudomonas aeruginosa.
Tebipenem is an orally available carbapenem antibiotic, shows broad-spectrum activity against Gram-positive and -negative bacteria, except for Pseudomonas aeruginosa.
Specifikationer
Kemiskt namn eller material Tebipenem
Fysisk form Solid
CAS 161715-21-5
Färg Light Yellow
Kvantitet 10 mg
Anteckningar om renhetsgrad Research
Molekylformel C16H21N3O4S2
För användning med (applikation) COVID-19-immunoregulation
Synonym LJC 11036
Löslighetsinformation DMSO : 33.33 mg/mL (86.91 mM; Need ultrasonic) ∣H2O : 7.14 mg/mL (18.62 mM; Need ultrasonic)
LEDER O=C(C(N1C2=O)=C(SC3CN(C4=NCCS4)C3)[C@H](C)[C@]1([H])[C@@]2([H])[C@H](O)C)O
Molekylvikt (g/mol) 383.49
Formel vikt 383.49
Procent renhet 98.0%