missing translation for 'onlineSavingsMsg'
Läs mer

Tebipenem Tetrahydrate, TRC

Produktkod. 30423242
Change view
Click to view available options
Kvantitet:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30423242 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30423242 Supplier Toronto Research Chemicals Supplier No. TRCT01284025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material Tebipenem Tetrahydrate
CAS 936010-57-0
Molekylformel C16H29N3O8S2
Kvantitet 25 mg
Synonym (4R,5S,6S)-3-[[1-(4,5-Dihydro-2-thiazolyl)-3-azetidinyl]thio]-6-[(1R)-1-hydroxyethyl]-4-methyl-7-oxo-1-azabicyclo[3.2.0]hept-2-ene-2-carboxylic Acid Tetrahydrate, [4R-[4α,5β,6β(R*)]]-3-[[1-(4,5-Dihydro-2-thiazolyl)-3-azetidinyl]thio]-6-(1-hydroxyethyl)-4-methyl-7-oxo-1-azabicyclo[3.2.0]hept-2-ene-2-carboxylic Acid Tetrahydrate, LJC 11036 Tetrahydrate
InChI formel InChI=1S/C16H21N3O4S2.4H2O/c1-7-11-10(8(2)20)14(21)19(11)12(15(22)23)13(7)25-9-5-18(6-9)16-17-3-4-24-16;;;;/h7-11,20H,3-6H2,1-2H3,(H,22,23);4*1H2/t7?,8?,10-,11-;;;;/m1..../s1
LEDER OC(C1=C(SC2CN(C3=NCCS3)C2)[C@H](C)[C@@]([C@@]4([H])[C@H](O)C)([H])N1C4=O)=O.O.O.O.O
IUPAC-namn (4R,5S,6S)-3-((1-(4,5-dihydrothiazol-2-yl)azetidin-3-yl)thio)-6-((R)-1-hydroxyethyl)-4-methyl-7-oxo-1-azabicyclo[3.2.0]hept-2-ene-2-carboxylic acid tetrahydrate
Molekylvikt (g/mol) 455.55
Formel vikt 455.14
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.