Läs mer
Tebutam, TRC
Produktkod. 30415743
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30415743 50 mg
1 options
Produktkod. 30415743 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT13623050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Tebutam
CAS 35256-85-0
Molekylformel C15 H23 N O
Kvantitet 50 mg
Synonym Propanamide','2,2-dimethyl-N-(1-methylethyl)-N-(phenylmethyl)- (9CI','ACI), 2,2-Dimethyl-N-(1-methylethyl)-N-(phenylmethyl)propanamide (ACI), Butam, Comodor, N-Benzyl-N-isopropylpivalamide, N-Benzyl-N-isopropyltrimethylacetamide, Tebutam
InChI formel InChI=1S/C15H23NO/c1-12(2)16(14(17)15(3,4)5)11-13-9-7-6-8-10-13/h6-10,12H,11H2,1-5H3
LEDER CC(C)N(Cc1ccccc1)C(=O)C(C)(C)C
IUPAC-namn N-benzyl-2,2-dimethyl-N-propan-2-ylpropanamide
Molekylvikt (g/mol) 233.35
Formel vikt 233.178