Läs mer
Tecloftalam, TRC
Produktkod. 30410041
Change view
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30410041 100 mg
30426609 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30410041 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT013700100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 76280-91-6
Molekylformel C14 H5 Cl6 N O3
Molekylvikt (g/mol) 447.91
Synonym Benzoic acid, 2,3,4,5-tetrachloro-6-[[(2,3-dichlorophenyl)amino]carbonyl]- (9CI, ACI), 2,3,4,5-Tetrachloro-6-[[(2,3-dichlorophenyl)amino]carbonyl]benzoic acid (ACI), 2,3,4,5-Tetrachloro-6-((2,3-dichlorophenyl)carbamoyl)benzoicacid, Shirahagen S, Techlofthalam, Tecloftalam
IUPAC-namn 2,3,4,5-tetrachloro-6-[(2,3-dichlorophenyl)carbamoyl]benzoic acid
LEDER OC(=O)c1c(Cl)c(Cl)c(Cl)c(Cl)c1C(=O)Nc2cccc(Cl)c2Cl
Specifikationer
Kemiskt namn eller material Tecloftalam
Kvantitet 100 mg
InChI formel InChI=1S/C14H5Cl6NO3/c15-4-2-1-3-5(8(4)16)21-13(22)6-7(14(23)24)10(18)12(20)11(19)9(6)17/h1-3H,(H,21,22)(H,23,24)
Formel vikt 444.8401