Läs mer
Juchheim Laborgeraete™ Teclu Safety Burner
Gastyp:
Naturgas
Propan
Höjd (metrisk):
150 mm
200 mm
230 mm
Beskrivning
- For operation with option of natural gas or propane gas
- With needle valve
- With DVGW certification
- With air control screen
- Particularly good stand due to heavy, high-quality material
Specifikationer
Specifikationer
| Produkttyp | Bunsen Burner |
| Gastyp | Naturgas |
| Material | Brass |
| Beskrivning | Gas consumption approx. 116 L/h (0.0813 kg/h), Heating capacity approx. 3398 kJ/h |
| Värmevärde | 3398 kJ/h |
| Höjd (metrisk) | 150 mm |
| Inkluderar | Needle Valve |