Läs mer
Tedizolid phosphate, MedChemExpress
Produktkod. 16486208
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
50 mg
100 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16486208 10 mg
16428807 5 mg
16406576 100 mg
16402347 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16486208 Leverantör Medchem Express Leverantörsnummer HY14855B
MedChemExpress Tedizolid phosphate (TR-701FA) is a novel oxazolidinone with activity against Gram-positive pathogens.
Tedizolid phosphate (TR-701FA) is a novel oxazolidinone with activity against Gram-positive pathogens.
Specifikationer
Kemiskt namn eller material Tedizolid phosphate
CAS 856867-55-5
Färg Off-White
Anteckningar om renhetsgrad Research
Molekylformel C17H16FN6O6P
Kvantitet 10 mg
Synonym TR-701FA
Löslighetsinformation DMSO : ≥ 36 mg/mL (79.94 mM) ∣H2O : 0.1 mg/mL (0.22 mM; Need ultrasonic)
LEDER O=C1O[C@@H](COP(O)(O)=O)CN1C2=CC=C(C3=CC=C(C4=NN(C)N=N4)N=C3)C(F)=C2
Molekylvikt (g/mol) 450.32
Formel vikt 450.32
Procent renhet 99.65%
Kvalitet Research
Fysisk form Powder
För användning med (applikation) COVID-19-immunoregulation