Läs mer
Tedizolid, TRC
Produktkod. 30004071
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30004071 50 mg
30012133 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30004071 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT01375050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 856866-72-3
Molekylformel C17 H15 F N6 O3
Molekylvikt (g/mol) 370.34
Synonym 2-Oxazolidinone, 3-[3-fluoro-4-[6-(2-methyl-2H-tetrazol-5-yl)-3-pyridinyl]phenyl]-5-(hydroxymethyl)-, (5R)-, (5R)-3-[3-Fluoro-4-[6-(2-methyl-2H-tetrazol-5-yl)-3-pyridinyl]phenyl]-5-(hydroxymethyl)-2-oxazolidinone, DA 7157, Sivextro, TR 700, Tedizolid
IUPAC-namn (5R)-3-[3-fluoro-4-[6-(2-methyltetrazol-5-yl)pyridin-3-yl]phenyl]-5-(hydroxymethyl)-1,3-oxazolidin-2-one
LEDER Cn1nnc(n1)c2ccc(cn2)c3ccc(cc3F)N4C[C@H](CO)OC4=O
Specifikationer
Kemiskt namn eller material Tedizolid
Kvantitet 50 mg
InChI formel InChI=1S/C17H15FN6O3/c1-23-21-16(20-22-23)15-5-2-10(7-19-15)13-4-3-11(6-14(13)18)24-8-12(9-25)27-17(24)26/h2-7,12,25H,8-9H2,1H3/t12-/m1/s1
Formel vikt 370.119