missing translation for 'onlineSavingsMsg'
Läs mer

Invitrogen™ TEF Polyclonal Antibody
GREENER_CHOICE

Produktkod. 16306904
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16306904 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 16306904 Leverantör Invitrogen™ Leverantörsnummer PA578195

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

Denna artikel kan inte returneras. Se returpolicy

Rabbit Polyclonal Antibody

Positive Control: TEF-transfected 293T Predicted Reactivity: Mouse (97%), Rat (98%), Sheep (95%), Rhesus Monkey (100%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

TEF (thyrotrophic embryonic factor), also known as KIAA1655, is a 303 amino acid nuclear transcription factor that belongs to the bZIP (basic region/leucine zipper) family and PAR (proline and acidic amino acid-rich) subfamily. TEF binds DNA as either a homodimer or heterodimer, and is known to transactivate the TSH beta promoter. While broadly expressed in adults, TEF is only found in developing embryonic anterior pituitary gland. TEF accumulates according to a robust circadian rhythm and has also been found to inhibit cell growth by downregulating beta chain expression of cytokine receptors. The functional domains of TEF are highly homologous with other members of the PAR-bZIP subfamily, including albumin D box-binding protein (DABP), human hepatic leukemia factor (HLF) and chicken vitellogenin gene-binding protein (VBP).
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen TEF
Användningsområden Western Blot
Klassificering Polyclonal
Koncentration 1.91 mg/mL
Konjugera Unconjugated
Formulering PBS with 20% glycerol and 0.01% thimerosal; pH 7
Gen TEF
Genaccessionsnr. Q10587
Gene Alias 2310028D20Rik; KIAA1655; TEF; TEF, PAR bZIP transcription factor; thyrotroph embryonic factor; thyrotrophic embryonic factor
Gensymboler TEF
Värd art Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human TEF. The exact sequence is proprietary.
Reningsmetod Antigen affinity chromatography
Kvantitet 100 μL
Regulatorisk status RUO
Primär eller sekundär Primary
Gen-ID (Entrez) 7008
Målarter Human
Innehåll och lagring Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Produkttyp Antibody
Form Liquid
Isotyp IgG
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.