Läs mer
Tefluthrin, TRC
Produktkod. 30041348
Change view
Kvantitet:
100 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30041348 1 g
18052430 100 mg
18060220 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30041348 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT0138001G
Denna vara kräver en prisuppdatering innan den beställs.
Begär ett pris för denna vara här.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 79538-32-2
Molekylformel C17 H14 Cl F7 O2
Molekylvikt (g/mol) 418.73
Synonym Cyclopropanecarboxylic acid, 3-(2-chloro-3,3,3-trifluoro-1-propenyl)-2,2-dimethyl-, (2,3,5,6-tetrafluoro-4-methylphenyl)methyl ester, [1α,3α(Z)]-(±)-, Cyclopropanecarboxylic acid, 3-[(1Z)-2-chloro-3,3,3-trifluoro-1-propenyl]-2,2-dimethyl-, (2,3,5,6-tetrafluoro-4-methylphenyl)methyl ester, (1R,3R)-rel- (9CI), A 14974, Cyclopropanecarboxylic acid, 3-(2-chloro-3,3,3-trifluoro-1-propenyl)-2,2-dimethyl-, (2,3,5,6-tetrafluoro-4-methylphenyl)methyl ester, [1α,3α(Z)]-, Force, Force (pesticide), Force 3G, Force CS, JF 6064, Komet RP, PP 993, ProShield, R 151993, Tefluthrin, Tefluthrin CS, Cyclopropanecarboxylic acid, 3-[(1Z)-2-chloro-3,3,3-trifluoro-1-propen-1-yl]-2,2-dimethyl-, (2,3,5,6-tetrafluoro-4-methylphenyl)methyl ester, (1R,3R)-rel-
IUPAC-namn (2,3,5,6-tetrafluoro-4-methylphenyl)methyl (1S,3S)-3-[(Z)-2-chloro-3,3,3-trifluoroprop-1-enyl]-2,2-dimethylcyclopropane-1-carboxylate
LEDER Cc1c(F)c(F)c(COC(=O)[C@H]2[C@@H](\C=C(/Cl)\C(F)(F)F)C2(C)C)c(F)c1F
Specifikationer
Kemiskt namn eller material Tefluthrin
Anteckningar om renhetsgrad HPLC
Kvantitet 1 g
InChI formel InChI=1S/C17H14ClF7O2/c1-6-11(19)13(21)7(14(22)12(6)20)5-27-15(26)10-8(16(10,2)3)4-9(18)17(23,24)25/h4,8,10H,5H2,1-3H3/b9-4-/t8-,10-/m1/s1
Formel vikt 418.0571
Procent renhet >95%