Läs mer
Tegafur, TRC
Produktkod. 30136450
Change view
Kvantitet:
5 mg
50 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136450 5 mg
30010137 50 mg
30006938 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30136450 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT0139005MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 17902-23-7
Molekylformel C8 H9 F N2 O3
Molekylvikt (g/mol) 200.17
Synonym 2,4(1H,3H)-Pyrimidinedione, 5-fluoro-1-(tetrahydro-2-furanyl)-, Uracil, 5-fluoro-1-(tetrahydro-2-furyl)- (8CI), 5-Fluoro-1-(tetrahydro-2-furanyl)-2,4(1H,3H)-pyrimidinedione, 1-(2-Tetrahydrofuryl)-5-fluorouracil, 1-(Tetrahydro-2-furanyl)-5-fluoro-2,4-pyrimidinedione, 1-(Tetrahydro-2-furanyl)-5-fluorouracil, 5-Fluoro-1-(2-tetrahydrofuranyl)uracil, 5-Fluoro-1-(tetrahydro-2-furanyl)-2,4-pyrimidinedione, 5-Fluoro-1-(tetrahydro-2-furyl)uracil, 5-Fluoro-1-(tetrahydro-2'-furyl)uracil, Citofur, Coparogin, Exonal, FT 207, Fental, Fluaid, Fluorafur, Fluorofur, Franrose, Ftorafur, Fulaid, Fulfeel, Furafluor, Furofutran, Futraful, Lamar, Lifril, MJF 12264, N1-(2-Tetrahydrofuranyl)-5-fluorouracil, N1-(2-Tetrahydrofuryl)-5-fluorouracil, N1-(2'-Tetrahydrofuryl)-5-fluorouracil, NSC 148958, Neberk, Nitobanil, Racemic Ftorafur, Riol, Sinoflurol, Sunfural, TS 1, Tefsiel C, Tegafur, Utefos
IUPAC-namn 5-fluoro-1-(oxolan-2-yl)pyrimidine-2,4-dione
LEDER FC1=CN(C2CCCO2)C(=O)NC1=O
Specifikationer
Kemiskt namn eller material Ftorafur
Anteckningar om renhetsgrad HPLC
Kvantitet 5 mg
InChI formel InChI=1S/C8H9FN2O3/c9-5-4-11(6-2-1-3-14-6)8(13)10-7(5)12/h4,6H,1-3H2,(H,10,12,13)
Formel vikt 200.0597
Procent renhet >95%