Läs mer
Tegaserod maleate, MedChemExpress
Produktkod. 16472117
Encompass_Preferred
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16472117 50 mg
16405978 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 16472117 Leverantör Medchem Express Leverantörsnummer HY14153A
MedChemExpress Tegaserod maleate is a selective 5-HT4 receptor partial agonist and a 5-HT2B receptor antagonist. Tegaserod maleate exhibits a promotile effect throughout the gastrointestinal (GI) tract.
Tegaserod maleate is a selective 5-HT4 receptor partial agonist and a 5-HT2B receptor antagonist. Tegaserod maleate exhibits a promotile effect throughout the gastrointestinal (GI) tract.
Specifikationer
Kemiskt namn eller material Tegaserod maleate
CAS 189188-57-6
Färg Light Yellow
Anteckningar om renhetsgrad Research
Molekylformel C20H27N5O5
Kvantitet 50 mg
Synonym SDZ-HTF-919 HTF-919
Löslighetsinformation DMSO : ≥ 35 mg/mL (83.84 mM) ∣H2O : 1 mg/mL (2.40 mM; Need ultrasonic)
LEDER O=C(O)/C=C\C(O)=O.COC1=CC=C(NC=C2/C=N/NC(NCCCCC)=N)C2=C1
Molekylvikt (g/mol) 417.46
Formel vikt 417.46
Procent renhet 99.75%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Neuroscience-Neuromodulation