missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TEM1 Mouse anti-Human, Clone: 1F9B4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 60170-1-IG-20UL
Additional Details : Weight : 0.00010kg
Description
CD248 (CD248 Molecule) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene include calcium ion binding and extracellular matrix binding. An important paralog of this gene is CD93. CD248 may play a role in tumor angiogenesis.Specifications
TEM1 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9HCU0 | |
CD248 | |
TEM1 Fusion Protein Ag13334 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Western Blot, Immunohistochemistry (Paraffin) | |
1F9B4 | |
Unconjugated | |
CD248 | |
CD164L1, CD248, CD248 molecule, endosialin, Endosialin, TEM1, Tumor endothelial marker 1 | |
Mouse | |
Caprylic Acid Precipitation | |
RUO | |
57124 | |
-20°C | |
Liquid |