Läs mer
Novus Biologicals™ TEM7/PLXDC1 Antibody Pair [HRP]
Produktkod. 18238656 Handla allt Bio Techne Produkter
Kvantitet:
15 Plates
5 tallrikar
Produktkod. 18238656
Brand: Novus Biologicals™ NBP27936315PLATES
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
Mouse Monoclonal Antibody Pair
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human TEM7/PLXDC1.
TRUSTED_SUSTAINABILITY
Specifikationer
Analysintervall 156.25 pg/mL
Analyskänslighet 156.25-10000 pg/mL
Analysera Sandwich ELISA
Konjugera HRP
Produkttyp Antibody Pairs
Specificitet This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human TEM7/PLXDC1.
Målarter Human
För användning med (applikation) Sandwich ELISA
Gene Alias FLJ45632, plexin domain containing 1,2410003I07Rik, plexin domain-containing protein 1, TEM3FLJ36270, TEM7DKFZp686F0937, Tumor endothelial marker 3, Tumor endothelial marker 7
Gen-ID (Entrez) 57125
Gensymbol PLXDC1
Värd art Mouse
Kit Innehåll
  • Detection Antibody: 0.5 mg/mL mouse anti-TEM7/PLXDC1 monoclonal antibody conjugated to horseradish-peroxidase (HRP).
  • Capture Antibody: 0.3 mg/mL of mouse anti-TEM7/PLXDC1 monoclonal antibody.
  • Standard: Each vial contains 270 ng of recombinant TEM7/PLXDC1.
  • Kvantitet 15 Plates
    Regulatorisk status RUO
    Forskningsdisciplin Angiogenesis
    Förvaringskrav Storage is content dependent.
    Mål TEM7/PLXDC1
    Teknik 1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in CBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 10 ng/mL is recommended.