Läs mer
Tembotrione, TRC
Produktkod. 30427317
Change view
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30427317 100 mg
30416922 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30427317 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT172533100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 335104-84-2
Molekylformel C17 H16 Cl F3 O6 S
Molekylvikt (g/mol) 440.82
Synonym 2-[2-Chloro-4-(methylsulfonyl)-3-[(2,2,2-trifluoroethoxy)methyl]benzoyl]-1,3-cyclohexanedione, BAY 747, Tembotrione
IUPAC-namn 2-[2-chloro-4-methylsulfonyl-3-(2,2,2-trifluoroethoxymethyl)benzoyl]cyclohexane-1,3-dione
LEDER CS(=O)(=O)c1ccc(C(=O)C2C(=O)CCCC2=O)c(Cl)c1COCC(F)(F)F
Specifikationer
Kemiskt namn eller material Tembotrione
Kvantitet 100 mg
InChI formel InChI=1S/C17H16ClF3O6S/c1-28(25,26)13-6-5-9(15(18)10(13)7-27-8-17(19,20)21)16(24)14-11(22)3-2-4-12(14)23/h5-6,14H,2-4,7-8H2,1H3
Formel vikt 440.0308