Läs mer
Temephos, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Temephos |
| CAS | 3383-96-8 |
| Molekylformel | C16 H20 O6 P2 S3 |
| Kvantitet | 100 mg |
| Synonym | Phosphorothioic acid','Op,Op'-(thiodi-4,1-phenylene) Op,Op,Op',Op'-tetramethyl ester, Phosphorothioic acid','O,O-dimethyl ester','O,O-diester with 4,4'-thiodiphenol (7CI), Phosphorothioic acid','O,O'-(thiodi-4,1-phenylene) O,O,O',O'-tetramethyl ester (9CI), Phosphorothioic acid','O,O'-(thiodi-p-phenylene) O,O,O',O'-tetramethyl ester (8CI), AC 52160, Abate, Abate 4E, Abathion, Azai-SS, Biothion, Biothon, Difenphos, Difos, Diphos, Diphos (pesticide), EI 52160, ENT 27165, Ecopro 1707, Experimental Insecticide 52,160, Lavifos, Mosq, Mosquit TB 10, Mosquit TB 100, Nephis, Nephis 1G, O,O,O',O'-Tetramethyl O,O'-thiodi-p-phenylene phosphorothioate, Procida, Temecal, Temefos, Temephos |
| InChI formel | InChI=1S/C16H20O6P2S3/c1-17-23(25,18-2)21-13-5-9-15(10-6-13)27-16-11-7-14(8-12-16)22-24(26,19-3)20-4/h5-12H,1-4H3 |
| LEDER | COP(=S)(OC)Oc1ccc(Sc2ccc(OP(=S)(OC)OC)cc2)cc1 |
| IUPAC-namn | [4-(4-dimethoxyphosphinothioyloxyphenyl)sulfanylphenoxy]-dimethoxy-sulfanylidene-λ5-phosphane |
| Molekylvikt (g/mol) | 466.47 |
| Formel vikt | 465.9897 |