Läs mer
Temocapril Hydrochloride, TRC
Produktkod. 30410696
Change view
Kvantitet:
5 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30410696 25 mg
30423502 50 mg
30415738 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30410696 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT01730125MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 110221-44-8
Molekylformel C23 H28 N2 O5 S2 . Cl H
Molekylvikt (g/mol) 513.07
IUPAC-namn 2-[(2S,6R)-6-[[(2S)-1-ethoxy-1-oxo-4-phenylbutan-2-yl]amino]-5-oxo-2-thiophen-2-yl-1,4-thiazepan-4-yl]acetic acid;hydrochloride
LEDER Cl.CCOC(=O)[C@H](CCc1ccccc1)N[C@H]2CS[C@@H](CN(CC(=O)O)C2=O)c3cccs3
Specifikationer
Kemiskt namn eller material Temocapril Hydrochloride
Kvantitet 25 mg
InChI formel InChI=1S/C23H28N2O5S2.ClH/c1-2-30-23(29)17(11-10-16-7-4-3-5-8-16)24-18-15-32-20(19-9-6-12-31-19)13-25(22(18)28)14-21(26)27;/h3-9,12,17-18,20,24H,2,10-11,13-15H2,1H3,(H,26,27);1H/t17-,18-,20-;/m0./s1
Formel vikt 512.1206