Läs mer
Temocaprilat, TRC
Produktkod. 30424067
Change view
Kvantitet:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30424067 250 mg
30414956 1 g
30421717 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30424067 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT017305250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 110221-53-9
Molekylformel C21 H24 N2 O5 S2
Molekylvikt (g/mol) 448.56
IUPAC-namn (2S)-2-[[(2S,6R)-4-(carboxymethyl)-5-oxo-2-thiophen-2-yl-1,4-thiazepan-6-yl]amino]-4-phenylbutanoic acid
LEDER OC(=O)CN1C[C@H](SC[C@H](N[C@@H](CCc2ccccc2)C(=O)O)C1=O)c3cccs3
Specifikationer
Kemiskt namn eller material Temocaprilat
Kvantitet 250 mg
InChI formel InChI=1S/C21H24N2O5S2/c24-19(25)12-23-11-18(17-7-4-10-29-17)30-13-16(20(23)26)22-15(21(27)28)9-8-14-5-2-1-3-6-14/h1-7,10,15-16,18,22H,8-9,11-13H2,(H,24,25)(H,27,28)/t15-,16-,18-/m0/s1
Formel vikt 448.1127